MilliporeSigma
  • Effects of glucose on contractile function, [Ca2+]i, and glycogen in isolated mouse skeletal muscle.
Effects of glucose on contractile function, [Ca2+]i, and glycogen in isolated mouse skeletal muscle.
American journal of physiology. Cell physiology (2002-05-09)
Ingrid Helander, Håkan Westerblad, Abram Katz
ABSTRACT
Extensor digitorum longus muscles were stimulated to contract to fatigue and allowed to recover for 2 h in the absence or presence of 5.5 or 11 mM extracellular glucose. This was followed by a second fatigue run, which ended when the absolute force was the same as at the end of the first run. During the first fatigue run, the fluorescence ratio for indo 1 increased [reflecting an increase in myoplasmic free Ca2+ concentration ([Ca2+]i)] during the initial tetani, peaking at approximately 115% of the first tetanic value, followed by a continuous decrease to approximately 90% at fatigue. During the first fatigue run, myofibrillar Ca2+ sensitivity was significantly decreased. During the second run, the number of tetani was 57 +/- 6% of initial force in muscles that recovered in the absence of glucose and 110 +/- 6 and 119 +/- 2% of initial force in muscles that recovered in 5.5 and 11 mM glucose, respectively. Fluorescence ratios during the first, peak, and last tetani did not differ significantly between the first and second fatigue runs during any of the three conditions. Glycogen decreased by almost 50% during the first fatigue run and did not change further after recovery in the absence of glucose. After recovery in the presence of 5.5 and 11 mM glucose, glycogen increased 32 and 42% above the nonstimulated control value (P < 0.01). These data demonstrate that extracellular glucose delays the decrease of tetanic force and [Ca2+]i during fatiguing stimulation and that glycogen supercompensation following contraction can occur in the absence of insulin.