MilliporeSigma
The catalytic asymmetric Mannich-type reactions in aqueous media.
Journal of the American Chemical Society (2002-05-16)
Shū Kobayashi, Tomoaki Hamada, Kei Manabe
ABSTRACT
The catalytic, diastereo- and enantioselective Mannich-type reaction of a hydrazono ester with silyl enol ethers in aqueous media has been successfully achieved with ZnF(2), a chiral diamine ligand, and trifluoromethanesulfonic acid.