MilliporeSigma
  • An in vitro oral biofilm model for comparing the efficacy of antimicrobial mouthrinses.
An in vitro oral biofilm model for comparing the efficacy of antimicrobial mouthrinses.
Caries research (2002-05-31)
S Shapiro, E Giertsen, B Guggenheim
ABSTRACT
The ability of commercial mouthrinses to reduce total viable counts of mixed microbial populations was examined using a previously developed in vitro model of supragingival plaque. Exploratory experiments aimed at fine-tuning the model indicated that optimal correspondence between in vitro and clinical results for chlorhexidine-containing formulations were obtained at a saliva:medium ratio of 70:30 (v/v); moreover, expanding the microbial population from 5 bacterial species to 5 bacterial species + Candida albicans had no noticeable impact on overall results. The efficacies of 12 different mouthrinse proprietary products containing chlorhexidine, hexetidine, octenidine, Triclosan, plant extracts, or aminefluoride/stannous fluoride vis-à-vis biofilm clearance were compared. All mouthrinses promoted a statistically significant reduction in microbial load compared to distilled water. The herbal- and phenolic-based products were substantially less effective than most chlorhexidine-containing mouthrinses, or mouthrinses containing hexetidine or octenidine. No significant difference between the plaque-clearing plaque-clearing abilities of Listerine and Meridol was observed. This polyspecies biofilm model can be a valuable tool for preclinical testing of antiplaque formulations, particularly during the product development stage.