MilliporeSigma
  • Asymmetric hydroformylation with highly crosslinked polystyrene-supported (R,S)-BINAPHOS-Rh(I) complexes: the effect of immobilization position.
Asymmetric hydroformylation with highly crosslinked polystyrene-supported (R,S)-BINAPHOS-Rh(I) complexes: the effect of immobilization position.
Bioorganic & medicinal chemistry letters (2002-06-28)
Fumitoshi Shibahara, Kyoko Nozaki, Takeshi Matsuo, Tamejiro Hiyama
ABSTRACT
A new class of polymer-supported (R,S)-BINAPHOS 1e in which the parent BINAPHOS has two alkoxy-substituents at the 3-positions of the phenyls, has been synthesised. Using its Rh(I) complex, asymmetric hydroformylation of olefins proceeded with higher enantioselectivity in some cases compared to the conventional polymer-supported 1c.
Sigma-Aldrich
cis-2-Butene, ≥99%
Sigma-Aldrich
trans-2-Butene, ≥99%
Sigma-Aldrich
2-Butene, mixture of cis and trans, ≥99%