MilliporeSigma
  • Inhibitory effects of natural sesquiterpenoids isolated from the rhizomes of Curcuma zedoaria on prostaglandin E2 and nitric oxide production.
Inhibitory effects of natural sesquiterpenoids isolated from the rhizomes of Curcuma zedoaria on prostaglandin E2 and nitric oxide production.
Planta medica (2002-07-03)
Chae Hee Hong, Min Soo Noh, Woo Young Lee, Sang Kook Lee
ABSTRACT
Beta-turmerone and ar-turmerone, sesquiterpenoids isolated from the rhizome of Curcuma zedoaria, inhibited lipopolysaccharide (LPS)-induced prostaglandin E 2 production in cultured mouse macrophage cell RAW 264.7 in a dose-dependent manner (IC 50 = 7.3 microM for beta-turmerone; IC 50 = 24.0 microM for ar-turmerone). In addition, these compounds exhibited inhibitory effects on LPS-induced nitric oxide production in the cell system.