MilliporeSigma
  • A new route to sulfonamides via intermolecular radical addition to pentafluorophenyl vinylsulfonate and subsequent aminolysis.
A new route to sulfonamides via intermolecular radical addition to pentafluorophenyl vinylsulfonate and subsequent aminolysis.
Organic letters (2002-07-19)
Stephen Caddick, Jonathan D Wilden, Hannah D Bush, Sjoerd N Wadman, Duncan B Judd
ABSTRACT
[reaction: see text] Various radical species generated from either the corresponding iodo- or bromo- compounds and tri-n-butyltin hydride were added in an intermolecular fashion to the activated acceptor pentafluorophenyl vinylsulfonate. The products of each reaction were then subjected to aminolysis with a variety of different amines.