MilliporeSigma
  • S-nitrosylation of matrix metalloproteinases: signaling pathway to neuronal cell death.
S-nitrosylation of matrix metalloproteinases: signaling pathway to neuronal cell death.
Science (New York, N.Y.) (2002-08-17)
Zezong Gu, Marcus Kaul, Boxu Yan, Steven J Kridel, Jiankun Cui, Alex Strongin, Jeffrey W Smith, Robert C Liddington, Stuart A Lipton
ABSTRACT
Matrix metalloproteinases (MMPs) are implicated in the pathogenesis of neurodegenerative diseases and stroke. However, the mechanism of MMP activation remains unclear. We report that MMP activation involves S-nitrosylation. During cerebral ischemia in vivo, MMP-9 colocalized with neuronal nitric oxide synthase. S-Nitrosylation activated MMP-9 in vitro and induced neuronal apoptosis. Mass spectrometry identified the active derivative of MMP-9, both in vitro and in vivo, as a stable sulfinic or sulfonic acid, whose formation was triggered by S-nitrosylation. These findings suggest a potential extracellular proteolysis pathway to neuronal cell death in which S-nitrosylation activates MMPs, and further oxidation results in a stable posttranslational modification with pathological activity.
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)