MilliporeSigma
  • On-resin real-time reaction monitoring of solid-phase oligosaccharide synthesis.
On-resin real-time reaction monitoring of solid-phase oligosaccharide synthesis.
Journal of the American Chemical Society (2002-10-24)
Shino Manabe, Yukishige Ito
ABSTRACT
On-resin real-time monitoring by a combination of a color test of (p-nitrobenzyl)pyridine and Disperse Red was developed for oligosaccharide synthesis.