MilliporeSigma
  • Inhibition of photosystem II of spinach by the respiration inhibitors piericidin A and thenoyltrifluoroacetone.
Inhibition of photosystem II of spinach by the respiration inhibitors piericidin A and thenoyltrifluoroacetone.
Bioscience, biotechnology, and biochemistry (2002-10-29)
Nobuhiro Ikezawa, Kentaro Ifuku, Tsuyoshi Endo, Fumihiko Sato
ABSTRACT
The effects of several respiration inhibitors on photosystem II (PS II) were investigated. Among the agents tested, piericidin A and thenoyltrifluoroacetone (TTFA) inhibited the photosynthetic electron transport of spinach as measured from chlorophyll (Chl) fluorescence parameters (Fm'-F)/Fm' and Fv/Fm. Using specific donors and acceptors of electrons, we identified the sites of inhibition in and around the PS II complex; the site of inhibition by TTFA was between QA, primary quinone acceptor in PS II, and QB, secondary quinone acceptor, in the acceptor side of P680, the reaction center Chl of PS II, while inhibition by piericidin A of the acceptor side was downstream of Q(B), out of the PS II complex. Both agents also inhibited the donor side of P680, probably between tyrosine-161 of the reaction center protein of PS II and P680.