MilliporeSigma
  • Valine 114 replacements in archaeal elongation factor 1 alpha enhanced its ability to interact with aminoacyl-tRNA and kirromycin.
Valine 114 replacements in archaeal elongation factor 1 alpha enhanced its ability to interact with aminoacyl-tRNA and kirromycin.
Biochemistry (2002-12-05)
Mariorosario Masullo, Piergiuseppe Cantiello, Barbara De Paola, Antonio Fiengo, Luigi Vitagliano, Adriana Zagari, Paolo Arcari
ABSTRACT
Valine 114 in the D(109)AAILVVA sequence of elongation factor 1alpha from the archaeon Sulfolobus solfataricus (SsEF-1alpha) was substituted with an acidic (V114E), basic (V114K), or cavity-forming (V114A) residue, and the effects on the biochemical properties of the factor were investigated. This sequence is well-conserved among most of eukaryal and eubacterial counterparts, and in the three-dimensional structure of SsEF-1alpha, V114 is located in a hydrophobic pocket near the first GDP-binding consensus sequence G(13)XXXXGK[T,S] [Vitagliano, L., Masullo, M., Sica, F., Zagari, A., and Bocchini, V. (2001) EMBO J. 20, 5305-5311]. These mutants displayed functions absent in the wild-type factor. In fact, although they exhibited a rate in poly(Phe) incorporation almost identical to that of SsEF-1alpha, V114K and V114A exhibited an affinity for GDP and GTP higher and a capability to bind heterologous aa-tRNA stronger than that elicited by SsEF-1alpha but similar to that of eubacterial EF-Tu. V114E instead displayed not only a weaker binding capability for aa-tRNA but also a lower affinity for GDP. The intrinsic GTPase activity of V114E was drastically reduced compared to those of SsEF-1alpha, V114K, and V114A. Interestingly, the decreased intrinsic GTPase activity of V114E was partially restored by kirromycin, an effect already observed for the G13A mutant of SsEF-1alpha [Masullo, M., Cantiello, P., de Paola, B., Catanzano, F., Arcari, P., and Bocchini, V. (2002) Biochemistry 41, 628-633]. Finally, the V114A substitution showed only a marginal effect on both the thermostability and thermophilicity of SsEF-1alpha, whereas V114K and V114E replacements strongly destabilized the molecule.