MilliporeSigma
Occupational asthma from fungicides fluazinam and chlorothalonil.
Occupational and environmental medicine (2002-12-25)
A Draper, P Cullinan, C Campbell, M Jones, A Newman Taylor
ABSTRACT
We report two cases of occupational asthma caused by sensitisation to powdered fungicides fluazinam and chlorothalonil, from the same fungicide formulation plant. Both developed work related lower respiratory symptoms after a latent interval of asymptomatic exposure. The diagnosis in each case was confirmed with a serial peak flow record in the workplace followed by specific inhalation tests. These fungicides are known to cause dermatitis; this report indicates that these compounds can induce specific immunological reactions in the airways as well as skin.
Product Description
Supelco
Fluazinam, PESTANAL®, analytical standard