MilliporeSigma
Kinetics of kavain and its metabolites after oral application.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-05-03)
F Tarbah, H Mahler, B Kardel, W Weinmann, D Hafner, Th Daldrup
ABSTRACT
Kavain metabolism in humans was the target of this current investigation. In the present study a high-performance liquid chromatographic (HPLC-DAD) assay method for the simultaneous determination of kavain and its main metabolites (p-hydroxykavain, p-hydroxy-5,6-dehydrokavain and p-hydroxy-7,8-dihydrokavain) in serum and urine was developed and validated. The metabolites were mainly excreted in the form of their conjugates. All kavain metabolites were detectable in serum and urine, except for p-hydroxy-7,8-dihydrokavain, which was found in urine only. Confirmation of the results and identification of the metabolites were performed by LC-MS or LC-MS-MS. Kinetics of kavain and its metabolites in serum were investigated after administration of a single oral dose (800 mg kavain). Within 1 and 4 h after uptake, the serum concentrations ranged between 40 and 10 ng/ml for kavain, 300 and 125 ng/ml for p-hydroxykavain, 90 and 40 ng/ml for o-desmethyl-hydroxy-5,6-dehydrokavain, and 50 and 30 ng/ml for 5,6-dehydrokavain.