MilliporeSigma
  • Influence of 1-aminocyclohexane-1-carboxylic acid in position 2 or 3 of AVP and its analogues on their pharmacological properties.
Influence of 1-aminocyclohexane-1-carboxylic acid in position 2 or 3 of AVP and its analogues on their pharmacological properties.
The journal of peptide research : official journal of the American Peptide Society (2003-06-26)
B Jastrzebska, I Derdowska, W Kowalczyk, A Machova, J Slaninová, B Lammek
ABSTRACT
In this study we describe the synthesis and some pharmacological properties of seven new analogues of arginine vasopressin (AVP) substituted in position 2 or 3 with 1-aminocyclohexane-1-carboxylic acid (Acc). All peptides were tested for the pressor, antidiuretic and uterotonic in vitro activities. The Acc3 modifications of AVP, dAVP, [d-Arg8]VP and [Cpa1]AVP have been found to be deleterious for interaction with all three neurohypophyseal hormone receptors, as judged from the several orders of magnitude decreased biological activities, whereas Acc2 substitution selectively altered the interaction with the receptors. Two of the new analogues, [Acc2]AVP and [Acc2, d-Arg8]AVP, are potent antidiuretic agonists. [Acc2]AVP exhibits moderate pressor agonistic activity and weak antiuterotonic properties. [Acc2, d-Arg8]AVP has been found to be a weak antagonist in the pressor and uterotonic tests. Another analogue - [Cpa1, Acc3]AVP - turned out to be a highly selective V2 agonist. This is an unexpected effect, as its parent peptide, [Cpa1]AVP is a very potent V1a receptor antagonist. This is the first Cpa1 modification to have resulted in V2 agonism enhancement. Besides providing useful information about structure-activity relationships, our results could open up new possibilities in the design of highly potent and selective V2 agonists.