MilliporeSigma
  • [Uniform design for the best conditions of processing heixi in shenrong heixi pills] off.
[Uniform design for the best conditions of processing heixi in shenrong heixi pills] off.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1992-06-01)
X Li, D Huang, G Huang, D Xi
ABSTRACT
Uniform design has been applied to select the optimum technological conditions for processing Heixi in Shenrong Heixi Pills, using the contents of the main constituent of Heixi SNS as the optimum target. The best technological conditions have thus been found.