MilliporeSigma
Biogeochemistry of fluoride in a plant-solution system.
Journal of environmental quality (2003-12-17)
C L Mackowiak, P R Grossl, B G Bugbee
ABSTRACT
Fluoride (F-) pollutants can harm plants and the animals feeding on them. However, it is largely unknown how complexing and chelating agents affect F bioavailability. Two studies were conducted that measured F- bioavailability and uptake by rice (Oryza sativa L.). In the first study, rice was grown in solution culture (pH 5.0) with 0, 2, or 4 mM F- as KF to compare the interaction of F- with humic acid (HA) and with a conventional chelating agent, N-hydroxyethylenthylenediaminetriacetic acid (HEDTA). In the second study, F was supplied at 0, 0.5, 1.0, and 2.0 mM KF with an additional 2 mM F- treatment containing solution Ca at 2x (2 mM Ca) the level used in the first study, to test the effect added Ca had on F- availability and uptake. Total biomass was greatest with HEDTA and F- < 1 mM. Leaf and stem F concentrations increased exponentially as solution F- increased linearly, with nearly no F partitioning into the seed. Results suggest that F was taken up as HF0 while F- uptake was likely restricted. Additionally, F- competed with HA for Ca, thus preventing the formation of Ca-HA flocculents. The addition of soluble Ca resulted in the precipitation of CaF2 solids on the root surface, as determined by tissue analysis and energy dispersive X-ray spectroscopy.
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, BioXtra, ≥98%
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, ~98%