MilliporeSigma
  • A prospective, randomised, cross-over study comparing the effects of clomiphene citrate and cyclofenil on endometrial morphology in the luteal phase of normal, fertile women.
A prospective, randomised, cross-over study comparing the effects of clomiphene citrate and cyclofenil on endometrial morphology in the luteal phase of normal, fertile women.
British journal of obstetrics and gynaecology (1992-12-01)
T C Li, M A Warren, C Murphy, S Sargeant, I D Cooke
ABSTRACT
To examine the effect of two anti-oestrogens, clomiphene citrate and cyclofenil, on endometrial morphology in the luteal phase. A prospective, randomised, cross-over study. Jessop Hospital for Women, Sheffield, UK. 10 women who were previously fertile and regularly cycling. The administration of clomiphene citrate or cyclofenil in the treatment cycles. A LH timed endometrial biopsy was taken on day LH + 6. Histological dating and morphometric analysis of the endometrial sample. Only one out of 10 subjects had abnormal endometrium. There was no difference in the results between cycles treated with clomiphene citrate and cyclofenil. Clomiphene citrate or cyclofenil does not have a major adverse effect on endometrial morphology in the luteal phase of normal fertile subjects. The possible adverse effects of anti-oestrogens on endometrium may have been previously overestimated.