MilliporeSigma
New potent and selective A1 adenosine receptor agonists.
Bioorganic & medicinal chemistry (2004-09-01)
Sally A Hutchinson, Stephen P Baker, Joel Linden, Peter J Scammells
ABSTRACT
Thiirane analogs of ENAdo have been synthesised and found to be extremely potent and selective A(1) adenosine receptor agonists.
Sigma-Aldrich
Ethylene sulfide, 98%