MilliporeSigma
  • A conventional route for the synthesis of new oxazolidin-2-one derivatives with beta-aminoalanines.
A conventional route for the synthesis of new oxazolidin-2-one derivatives with beta-aminoalanines.
Chemical & pharmaceutical bulletin (2004-10-07)
Fumiko Fujisaki, Nobuhiro Abe, Kunihiro Sumoto
ABSTRACT
A conventional new route to the novel oxazolidin-2-one derivatives (3a-f) having two substituents on N-3 and C-4 in the oxazolidin-2-one ring was established with racemic beta-aminoalanine derivatives (1) as the key starting materials.
Sigma-Aldrich
DL-2,3-Diaminopropionic acid monohydrochloride, 98%