MilliporeSigma
  • Isethionic acid and floridoside isolated from the red alga, Grateloupia turuturu, inhibit settlement of Balanus amphitrite cyprid larvae.
Isethionic acid and floridoside isolated from the red alga, Grateloupia turuturu, inhibit settlement of Balanus amphitrite cyprid larvae.
Biofouling (2004-11-17)
Claire Hellio, Christelle Simon-Colin, Anthony S Clare, Eric Deslandes
ABSTRACT
Isethionic acid (2-hydroxyethane sulfonic acid) and floridoside (2-O-alpha-D-galactopyranosylglycerol) were extracted from the red alga, Grateloupia turuturu, and tested for anti-settlement activity against cyprid larvae of the tropical barnacle, Balanus amphitrite and for their toxicity to nauplius larvae. Isethionic acid was active for anti-settlement but had the disadvantage of being toxic to nauplius larvae. Floridoside was a potent inhibitor of cyprid settlement at non-toxic concentrations to nauplii (0.01 mg ml(-1)).