MilliporeSigma
  • Inhibition of Leishmania donovani promastigote DNA topoisomerase I and human monocyte DNA topoisomerases I and II by antimonial drugs and classical antitopoisomerase agents.
Inhibition of Leishmania donovani promastigote DNA topoisomerase I and human monocyte DNA topoisomerases I and II by antimonial drugs and classical antitopoisomerase agents.
The Journal of parasitology (2004-11-26)
John Walker, Nancy G Saravia
ABSTRACT
We have compared the inhibitor sensitivities of DNA topoisomerase I (TOPI) from Leishmania donovani promastigotes and TOPs I and II of human monocytes using pentavalent and trivalent antimonials (SbV, SbIII) and classical TOP inhibitors. Bis-benzimidazoles (Hoechst-33258 and -33342) were potent inhibitors of both parasite and human TOPI, but Hoechst-33342 was markedly less cytotoxic to promastigotes than to monocytes in vitro. Leishmania donovani was also considerably less sensitive than monocytes to camptothecin, both at enzyme and cellular levels. Sodium stibogluconate (SSG) was the only antimonial to inhibit TOPI, exhibiting a significant (P < 0.05) 3-fold greater potency against the L. donovani enzyme but showed low cytotoxicities against intact promastigotes. The SbV meglumine antimoniate failed to inhibit TOPI and showed negligible cytotoxicities, whereas SbIII drugs were lethal to parasites and monocytes yet poor inhibitors of TOPI. Monocyte TOPII was inhibited by bis-benzimidazoles and insensitive to antimonials and camptothecin. The disparity between the high leishmanicidal activity and low anti-TOPI potency of SbIII indicates that in vivo targeting of L. donovani TOPI by the reductive pathway of antimonial activation is improbable. Nevertheless, the potent direct inhibition of TOPI by SSG and the differential interactions of camptothecin with L. donovani and human TOPI support the possibility of developing parasite-specific derivatives.
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, puriss., meets analytical specification of USP, 99.0-103.0%, powder
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, purum p.a., 99.0-103% (RT)
Sigma-Aldrich
Potassium antimonyl tartrate trihydrate, ACS reagent, ≥99%