MilliporeSigma
  • Gas chromatographic method for the analysis of allelopathic natural products in rye (Secale cereale L.).
Gas chromatographic method for the analysis of allelopathic natural products in rye (Secale cereale L.).
Journal of chromatography. A (2005-03-30)
M M Finney, D A Danehower, J D Burton
ABSTRACT
Accurate and reproducible methods for the analysis of plant allelochemicals are a requirement for the study of chemical interactions between plants. This paper describes a method for sample preparation and quantitative analysis of the allelopathic chemical content of rye (Secale cereale L.) using gas chromatography (GC). Sample preparation consists of extraction of freeze-dried rye vegetative tissue with aqueous ethanol followed by partitioning of the allelochemicals into ethyl acetate, evaporation, and derivatization using the trimethylsilylating reagent N-methyl-N-trimethylsilyltrifluoroacetamide. GC analysis of the silylated mixture was performed using flame ionization detection. This method permits analysis of all known rye allelopathic agents including 2,4-dihydroxy-1,4-benzoxazin-3-one, its corresponding glucoside, 2-benzoxazolinone, beta-hydroxybutyric acid, and beta-phenyllactic acid. Identities of all compounds were confirmed by GC/MS analysis.
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%