MilliporeSigma
  • Electronic and infrared spectral and thermal studies on the molecular complex of dibenzo-18-crown-6 and iodine.
Electronic and infrared spectral and thermal studies on the molecular complex of dibenzo-18-crown-6 and iodine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-05-03)
L A Shahada
ABSTRACT
The interaction of the crown ether dibenzo-18-crown-6 (DBC) with iodine has been studied in CHCl3 at room temperature. The charge-transfer absorptions, far infrared and thermal measurements of the formed charge-transfer complex were recorded and discussed. The results obtained show the formation of the pentaiodide complex with the general formula [(DBC)]+ I5-. The pentaiodide ion, I5-, is described as I3-(I2) confirmed by the observation of the characteristic absorptions for I3- ion around 365 and 290 nm. In addition, the far infrared spectrum of the solid complex shows the three vibrations of I3- unit is at 141, 113 and 71 cm(-1) assigned to nu(as)(I-I) and nu(s)(I-I) and delta(I3-), respectively, while the band related to the vibration of I2 unit is observed at 180 cm(-1). Vibration analysis of the obtained data shows that the symmetry of I3- unit could be non-linear with C2v symmetry. The structure of the formed pentaiodide complex was further supported by thermal gravimetric analysis measurements.
Sigma-Aldrich
Dibenzo-18-crown-6, 98%