MilliporeSigma
  • DNA hypomethylation induced by drinking water disinfection by-products in mouse and rat kidney.
DNA hypomethylation induced by drinking water disinfection by-products in mouse and rat kidney.
Toxicological sciences : an official journal of the Society of Toxicology (2005-07-15)
Lianhui Tao, Wei Wang, Long Li, Paula K Kramer, Michael A Pereira
ABSTRACT
Bromodichloromethane (BDCM), chloroform, dibromoacetic acid (DBA), dichloroacetic acid (DCA), and trichloroacetic acid (TCA) are chlorine disinfection by-products (DBPs) found in drinking water that have indicated renal carcinogenic and/or tumor promoting activity. We have reported that the DBPs caused DNA hypomethylation in mouse liver, which correlated with their carcinogenic and tumor promoting activity. In this study, we determined their ability to cause renal DNA hypomethylation. B6C3F1 mice were administered DCA or TCA concurrently with/without chloroform in their drinking water for 7 days. In male, but not female mouse kidney, DCA, TCA, and to a lesser extent, chloroform decreased the methylation of DNA and the c-myc gene. Coadministering chloroform increased DCA but not TCA-induced DNA hypomethylation. DBA and BDCM caused renal DNA hypomethylation in both male B6C3F1 mice and Fischer 344 rats. We have reported that, in mouse liver, methionine prevented DCA- and TCA-induced hypomethylation of the c-myc gene. To determine whether it would also prevent hypomethylation in the kidneys, male mice were administered methionine in their diet concurrently with DCA or TCA in their drinking water. Methionine prevented both DCA- and TCA-induced hypomethylation of the c-myc gene. The ability of the DBPs to cause hypomethylation of DNA and of the c-myc gene correlated with their carcinogenic and tumor promoting activity in mouse and rat kidney, which should be taken into consideration as part of their risk assessment. That methionine prevents DCA- and TCA-induced hypomethylation of the c-myc gene would suggest it could prevent their carcinogenic activity in the kidney.
Product Description
Supelco
Dibromoacetic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Dibromoacetic acid, 97%
Pricing and availability is not currently available.