MilliporeSigma
  • The first crystal structure of a family 31 carbohydrate-binding module with affinity to beta-1,3-xylan.
The first crystal structure of a family 31 carbohydrate-binding module with affinity to beta-1,3-xylan.
FEBS letters (2005-08-03)
Hiroshi Hashimoto, Youichi Tamai, Fumiyoshi Okazaki, Yutaka Tamaru, Toshiyuki Shimizu, Toshiyoshi Araki, Mamoru Sato
ABSTRACT
Here, we present the crystal structure of the family 31 carbohydrate-binding module (CBM) of beta-1,3-xylanase from Alcaligenes sp. strain XY-234 (AlcCBM31) determined at a resolution of 1.25A. The AlcCBM31 shows affinity with only beta-1,3-xylan. The AlcCBM31 molecule makes a beta-sandwich structure composed of eight beta-strands with a typical immunoglobulin fold and contains two intra-molecular disulfide bonds. The folding topology of AlcCBM31 differs from that of the large majority of other CBMs, in which eight beta-strands comprise a beta-sandwich structure with a typical jelly-roll fold. AlcCBM31 shows structural similarity with CBM structures of family 34 and family 9, which also adopt structures based on immunoglobulin folds.