MilliporeSigma
  • Phenolsulfonphthalein transport by potential-sensitive urate transport system.
Phenolsulfonphthalein transport by potential-sensitive urate transport system.
European journal of pharmacology (2005-08-09)
Shirou Itagaki, Soji Shimamoto, Mitsuru Sugawara, Michiya Kobayashi, Katsumi Miyazaki, Takeshi Hirano, Ken Iseki
ABSTRACT
The purpose of this study was to elucidate the transporter-mediated secretion systems for phenolsulfonphthalein in brush-border membranes. In human and rat renal brush-border membranes, a potential-sensitive transport system has been shown to be involved in the efflux of organic anions. The uptake of phenolsulfonphthalein into rat renal brush-border membrane vesicles was stimulated by an inside-positive membrane potential. This potential-sensitive uptake of phenolsulfonphthalein was inhibited by probenecid, pyrazinoate and urate. p-Aminohippurate had no effect on the potential-sensitive uptake of phenolsulfonphthalein. Moreover, urate competitively inhibited the uptake of phenolsulfonphthalein. On the other hand, the uptake of phenolsulfonphthalein was slightly increased in the presence of an outward Cl- gradient. These results suggest that phenolsulfonphthalein has high affinity for the potential-sensitive urate transport system but has low affinity for an anion exchanger.
Sigma-Aldrich
Pyrazinecarboxylic acid, 99%