MilliporeSigma
Antiplatelet property of Curcuma longa L. rhizome-derived ar-turmerone.
Bioresource technology (2005-08-23)
H S Lee
ABSTRACT
The antiplatelet activities of Curcuma longa L. rhizome-derived materials were measured using a platelet aggregometer and compared with those of aspirin as antiplatelet agent. The active constituent from the rhizome of Curcuma longa L. was isolated and characterized as ar-turmerone by various spectral analyses. At 50% inhibitory concentration (IC50) value, ar-turmerone was effective in inhibiting platelet aggregation induced by collagen (IC50, 14.4 microM) and arachidonic acid (IC50, 43.6 microM). However, ar-turmerone had no effect on platelet activating factor or thrombin induced platelet aggregation. In comparison, ar-turmerone was significantly more potent platelet inhibitor than aspirin against platelet aggregation induced by collagen. These results suggested that ar-turmerone could be useful as a lead compound for inhibiting platelet aggregation induced by collagen and arachidonic acid.