MilliporeSigma
  • Iso-migrastatin congeners from Streptomyces platensis and generation of a glutarimide polyketide library featuring the dorrigocin, lactimidomycin, migrastatin, and NK30424 scaffolds.
Iso-migrastatin congeners from Streptomyces platensis and generation of a glutarimide polyketide library featuring the dorrigocin, lactimidomycin, migrastatin, and NK30424 scaffolds.
Journal of the American Chemical Society (2005-08-25)
Jianhua Ju, Si-Kyu Lim, Hui Jiang, Jeong-Woo Seo, Ben Shen
ABSTRACT
Iso-Migrastatin (10) has been shown to be the main natural product of Streptomyces platensis, which undergoes a facile, H2O-mediated rearrangement into dorrigocin A (2), 13-epi-dorrigocin A (11), dorrigocin B (3), and migrastatin (1). Eight new congeners (12-19) of 10 were characterized. They can undergo the same H2O-mediated rearrangement into the corresponding 1, 2, 3, and 11 analogues (20-43) or 1,4-Michael addition with cysteine to afford the corresponding analogues (44-51) of NK30424 A and B (5, 6). This study generated a 47-member library of glutarimide polyketides, setting the stage to investigate the SAR for this family of natural products. These results also established the absolute stereochemistry of 5 and 6 and shed new light into the post-polyketide synthase steps for 10 biosynthesis.
Sigma-Aldrich
Glutarimide, 98%