MilliporeSigma
Vibrational spectroscopy of N-phenylmaleimide.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-09-15)
Stewart F Parker
ABSTRACT
A combination of infrared, Raman and inelastic neutron scattering (INS) spectroscopies with density functional theory (DFT) calculations is used to provide a complete assignment of the vibrational spectra of N-phenylmaleimide and N-(perdeuterophenyl)maleimide. DFT is shown to give very good results for the frequencies and atomic displacements in the modes. These are used to generate INS spectra which are excellent agreement with the observed. The calculated infrared and Raman spectra are much less reliable, although this may be more of a presentation problem than a real failing.
Sigma-Aldrich
N-Phenylmaleimide, 97%