MilliporeSigma
  • Urinary ortho-tyrosine excretion in diabetes mellitus and renal failure: evidence for hydroxyl radical production.
Urinary ortho-tyrosine excretion in diabetes mellitus and renal failure: evidence for hydroxyl radical production.
Kidney international (2005-10-14)
Gergõ A Molnár, Zoltán Wagner, Lajos Markó, Tamás Kó Szegi, Márton Mohás, Béla Kocsis, Zoltán Matus, László Wagner, Mónika Tamaskó, István Mazák, Boglárka Laczy, Judit Nagy, István Wittmann
ABSTRACT
Phenylalanine is converted to para- and ortho-tyrosine by hydroxyl free radical, or to para-tyrosine by the phenylalanine hydroxylase enzyme. The aim of this study was to measure para- and ortho-tyrosine in the urine and plasma of patients with chronic renal disease and/or diabetes, to obtain information on the renal handling of the different tyrosine isomers and, furthermore, to measure urinary levels of 8-epi-prostaglandin-F(2alpha), a marker of lipid peroxidation. In our cross-sectional study we measured para-, ortho-tyrosine, and phenylalanine levels, using high performance liquid chromatography and 8-epi-prostaglandin-F(2alpha) with enzyme-linked immunosorbent assay (ELISA). We compared 4 groups: (1) controls (CONTR, N = 14), (2) patients with chronic kidney disease (CKD, N = 12), (3) patients with type 2 diabetes mellitus (DIAB, N = 17), (4) patients with chronic kidney disease and type 2 diabetes (DIAB-CKD, N = 19). We found a decreased plasma para-tyrosine level and decreased urinary para-tyrosine excretion in CKD patients, while the fractional excretion of para-tyrosine was similar in all 4 groups, approximately 1%. There was no difference in the plasma ortho-tyrosine levels between the groups. However, urinary ortho-tyrosine excretion was higher in all 3 groups of patients than in the CONTR group, and higher in DIAB and in DIAB-CKD patients than in CKD patients. The fractional excretion of ortho-tyrosine was significantly higher in DIAB and in DIAB-CKD patients than in the CONTR group. The fractional excretion of ortho-tyrosine exceeded 100% in the 2 diabetic groups. Urinary 8-epi-prostaglandin-F(2alpha)/creatinine ratio did not correlate with urinary ortho-tyrosine excretion. The difference between para-tyrosine levels of the groups is probably due to renal impairment, while there is indirect evidence for an increased tubular secretion or production of ortho-tyrosine in the kidney in diabetic patients with or without CKD.
Sigma-Aldrich
DL-o-Tyrosine, ≥96.0% (NT)
Pricing and availability is not currently available.