MilliporeSigma
  • Effect of a mixture of iprobenfos and malathion on the development of malathion resistance in the mosquito Culex pipiens pallens Coq.
Effect of a mixture of iprobenfos and malathion on the development of malathion resistance in the mosquito Culex pipiens pallens Coq.
Pest management science (2005-10-20)
Li-Ming Tao, Jian-Zhong Yang, Pei-Jun Zhuang, Zhen-Hua Tang
ABSTRACT
A malathion-resistant (RM) strain of Culex pipiens pallens Coq was obtained by successively selecting a field population with malathion in the laboratory. The synergistic effect of iprobenfos on malathion toxicity and alpha-naphthyl acetate (alpha-NA) esterase assay revealed that malathion resistance in the RM strain was associated with increased alpha-NA esterase activity and the synergism was mainly due to the inhibition by iprobenfos of this activity. There was no difference in alpha-NA esterase activity between the larvae and female adults in the susceptible (S) strain, but the activity in the adults was 13-fold higher than in the larvae of the RM strain. To understand the effect of the application of a mixture of iprobenfos and malathion on the evolution of malathion resistance, an artificial strain (Syn) was generated by mixing the RM and S strains with 0.1 frequency of the malathion-resistant individuals. The offspring of the Syn strain were divided into two sub-strains, Rm and Rm+ibp, which were successively treated with, respectively, malathion alone and malathion + iprobenfos (1:2) at LC70. In the mixture, the fungicide iprobenfos acted as a synergist of malathion. After treatment for 10 generations, the resistance level to malathion was 317.4-fold for the Rm sub-strain, whereas for the Rm+ibp sub-strain it was only 38.9-fold, compared with the Syn strain. Similar results were obtained by measurement of alpha-NA esterase activity from both larvae and female adults. The alpha-NA esterase activities in larvae and female adults at F10 generation were 2.6- and 10.9-fold from the Rm+ibp sub-strain and 5.7- and 98.5-fold from the Rm sub-strain, respectively, compared with the Syn strain. The above results suggested that iprobenfos, although it cannot completely stop or prevent the onset of malathion resistance, could dramatically delay its evolution.
Product Description
Supelco
Iprobenfos, PESTANAL®, analytical standard