MilliporeSigma
Enrichment and isolation of anaerobic hydrocarbon-degrading bacteria.
Methods in enzymology (2005-11-02)
Irene A Davidova, Joseph M Suflita
ABSTRACT
Recent progress in microbiology resulted in the enrichment and isolation of anaerobic bacteria capable of the biodegradation of various hydrocarbons under a variety of electron-accepting conditions. Problems challenging the enrichment and isolation of anaerobic hydrocarbonclastic organisms required new approaches and modifications of conventional microbiological techniques. This chapter summarizes the collective experience accumulated in this area starting from anaerobic sampling precautions and includes all stages of cultivation from the construction of initial incubations to final isolation steps and the evaluation of culture purity.
Sigma-Aldrich
2,2,4,4,6,8,8-Heptamethylnonane, 98%