MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Determination of some polycyclic aromatic hydrocarbons in filter tar of Turkish cigarettes.
Determination of some polycyclic aromatic hydrocarbons in filter tar of Turkish cigarettes.
Journal of separation science (2005-12-14)
Seker Fatma Aygün
ABSTRACT
A simultaneous determination method for the analysis of benzo[a]pyrene (BaP) and dibenzo[a,h]anthracene (DahA) in the filter tar of Turkish cigarettes has been developed. The method involved (a) the extraction of BaP and DahA with n-hexane from ACN solution in which the cigarette filters were extracted, and then (b) purification of the n-hexane extracts by elution on an XAD-2 column using n-hexane/dichloromethane (9:1, v/v) mixture. Separation and quantitative determination of BaP and DahA in the extracts were carried out by HPLC and fluorescence detection on a C18 RP column. The calculated recoveries for BaP and DahA were found in the range of 90-100% for each extraction and clean-up steps. Analysis of various filter tar of Turkish cigarettes showed that an average of 74.28 ng/filter of BaP and 5.24 ng/filter of DahA were present in Turkish cigarettes.
Product Description
Dibenz[a,h]anthracene, BCR®, certified reference material
Supelco
Dibenz[a,h]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland