- Antialgal, antibacterial and antifungal activity of two metabolites produced and excreted by cyanobacteria during growth.
Antialgal, antibacterial and antifungal activity of two metabolites produced and excreted by cyanobacteria during growth.
Microbiological research (2006-01-24)
Rainer-B Volk, Franz H Furkert
PMID16427523
ABSTRACT
The antimicrobial activity of two cyanobacterial exometabolites, norharmane (9H-pyrido(3,4-b)indole) and 4,4'-dihydroxybiphenyl, was determined in suspension assays. Good anticyanobacterial activities (concentrations of 8-80 microg ml(-1)) and moderate antibacterial (16-160 microg ml(-1)) and antifungal (32-40 microg ml(-1)) activities were found for both compounds. The natural function as allelopathic chemicals and a possible applicability as antifouling agents or leads for the development of new antifouling chemicals are discussed.