MilliporeSigma
  • Synthesis of diacetoxy acetal derivatives of santonin and their enhancing effects on HL-60 leukemia cell differentiation.
Synthesis of diacetoxy acetal derivatives of santonin and their enhancing effects on HL-60 leukemia cell differentiation.
Archives of pharmacal research (2006-02-24)
Seung Hyun Kim, Sun Young Chung, Tae Sung Kim, Bo Gil Choi
ABSTRACT
Several diacetoxy acetal analogues have been synthesized from santonin and assessed for their ability of inducing or enhancing the differentiation of human HL-60 leukemia cells. The compounds themselves had little effect on HL-60 cell differentiation. However, three analogues, 2a, 3a, and 5b, synergistically enhanced 1,25-dihydroxyvitamin D3 [1,25-(OH)2D3]-induced HL-60 cell differentiation when combined with 5 nM of dihydroxyvitamin D3 [1,25-(OH)2D3], a well-known differentiation inducer. Especially, the compound 5b profoundly enhanced the 1,25-(OH)2D3]-induced HL-60 cell differentiation.