MilliporeSigma
  • Metabolism of uniconazole-P in water-sediment systems under illumination.
Metabolism of uniconazole-P in water-sediment systems under illumination.
Environmental toxicology and chemistry (2006-03-08)
Rika Kodaka, Terumi Sugano, Toshiyuki Katagi
ABSTRACT
Aerobic soil metabolism of uniconazole-P ([S]-E-1-[4-chlorophenyl]-4,4-dimethyl-2-[1,2,4-triazole-1-yl]-penten-3-ol) and the effect of illumination on metabolic profiles were studied in the water-sediment system when spiked to water. Uniconazole-P was gradually partitioned to the sediment with an aquatic half-life of 6.9 d in darkness with formation of bound residues. Illumination of the system from a xenon lamp (>290 nm) greatly accelerated the degradation of uniconazole-P via photoinduced isomerization between E- and Z-isomers with a subsequent intramolecular cyclization, and its aquatic half-life was greatly reduced to 0.6 d. Kinetic analysis based on compartment models suggested the possible contribution of photodegradation at the water-sediment interface, leading to more formation of the cyclized derivative in the sediment.
Product Description
Supelco
Uniconazole, PESTANAL®, analytical standard