MilliporeSigma
  • Attenuation of contractility in rat epididymal vas deferens by Rho kinase inhibitors.
Attenuation of contractility in rat epididymal vas deferens by Rho kinase inhibitors.
Autonomic & autacoid pharmacology (2006-03-24)
N I B Amobi, I-P Chung, I C H Smith
ABSTRACT
The involvement of Ca(2+) sensitization mediated through Rho kinase in the contractility of rat epididymal vas deferens was investigated using Rho kinase inhibitors, trans-4-[(1R)-1-aminoethyl]-N-4-pyridinilcyclohexanecarboxamide dihydrochloride (Y-27632) and 1-(5-isoquinolinesulphonyl)homopiperazine (HA 1077), in comparison with myosin light chain kinase (MLCK) inhibitors, wortmannin and 1-(5-chloronaphthalenesulphonyl)homopiperazine (ML-9) and agents that affect protein kinase C (PKC) and non-receptor tyrosine kinase intracellular signalling. 2 In Ca(2+)-free/ethyleneglycol-bis-(beta-aminoethylether)N,N,N('),N(')-tetraacetic acid (EGTA) (1 mM) medium, noradrenaline evoked sustained contractions. Y-27632 and HA 1077 caused a concentration-dependent inhibition and complete relaxation (IC(50), 1.08 and 1.75 microM respectively). The Ca(2+)-free contraction was reduced by wortmannin (10 microM) or ML-9 (10 microM) but not by inhibitors of diacylglycerol metabolism, 3-[2-[4[bis(4-Fluoropheny)methylene]-1-piperidinyl]-2,3-dihydro-2-thioxi-4(H)-quinazolinone (R59949) (10 microm) or 1,6-bis(cyclohexyloximinocarbonylamino)hexane (RHC-80267) (10 microM) or by the phospholipase A(2) (PLA(2)) inhibitor, quinacrine (up to 100 microM) or tyrosine kinase inhibitor, genistein (30 microM). 3 In the presence of Ca(2+) (2.5 mM), noradrenaline (100 microM) evoked rhythmic activity and biphasic tonic contractions. Y-27632 (1-10 microM) or HA 1077 (1-10 microM) reduced the amplitude of rhythmic activity and tonic contractions. ML-9 (10 microM) attenuated the occurrence of rhythmic activity and modestly reduced the tonic contractions. ML-9 (10 microM) combined with Y-27632 (10 microM) significantly reduced the tonic contractions. ML-9 (30 microM) alone (or combined with Y-27632 10 microM) suppressed the rhythmic activity and substantially reduced (or abolished) the tonic contractions. 4 Contractions evoked by high [K(+)](o) (120 mM) or alpha,beta-methylene ATP (10 microM) were reduced significantly by Y-27632 (1-3 microM) indicating that the Rho kinase signalling pathway is activated by direct tissue depolarization or by stimulation of ligand-gated P(2X) purinoceptors. 5 Collectively, these results indicate that Ca(2+)-sensitization mediated by Rho kinase is involved in agonist- or depolarization-induced contraction of rat epididymal vas deferens. It is the major contractile mechanism underlying noradrenaline-induced Ca(2+)-free responses. It contributes to Ca(2+)-dependent rhythmic contractility and optimizes the development of full contractile tension triggered through calmodulin/MLCK activation by stimulated influx of Ca(2+).