MilliporeSigma
On the nitrile effect in L-rhamnopyranosylation.
Carbohydrate research (2006-04-29)
David Crich, Mitesh Patel
ABSTRACT
It is shown that the use of 5% acetonitrile or propionitrile in dichloromethane functions to increase the beta-selectivity of a number of L-rhamnopyranosylation reactions conducted by the thioglycoside method with activation by the 1-benzenesulfinyl piperidine/trifluoromethanesulfonic anhydride couple. The use of more significant quantities of acetonitrile or propionitrile results in the formation of complex reaction mixtures containing little coupled product, but from which Ritter-type products can be isolated.
Sigma-Aldrich
Propionitrile, 99%
Supelco
Propionitrile, analytical standard
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)