MilliporeSigma
Precision study on capillary electrophoresis methods for metacycline.
Electrophoresis (2006-05-24)
Thao Do Thi, Romeo Pomponio, Roberto Gotti, Jan Saevels, Ben Van Hove, Willy Van Ael, Nele Matthijs, Yvan Vander Heyden, Roland Marini Djan'geing'a, Patrice Chiap, Philippe Hubert, Jacques Crommen, Huguette Fabre, Pieter Dehouck, Jos Hoogmartens, Ann Van Schepdael
ABSTRACT
A CE method for metacycline (MTC) determination was investigated in an inter-laboratory experiment. Many problems were encountered in this study, most of which were related to the transfer of the method to different CE equipment. The reported problems could be classified into different categories: problems related to the precision, to the parameters in the protocol, and to the MTC peak shape. As the peak shape problem was partially responsible for the poor precision, a new CE method was developed in order to obtain a good MTC peak shape on all equipment. The precision of this new method for MTC determination was examined in an intermediate precision study, where the influence of the factors "time" and "equipment" was investigated. Although the new method could be transferred to different instruments, the precision remained poor mainly due to the contributions of the between-replicate and the between-injection variances.