MilliporeSigma
  • High performance liquid chromatography analysis of 2-mercaptoethylamine (cysteamine) in biological samples by derivatization with N-(1-pyrenyl) maleimide (NPM) using fluorescence detection.
High performance liquid chromatography analysis of 2-mercaptoethylamine (cysteamine) in biological samples by derivatization with N-(1-pyrenyl) maleimide (NPM) using fluorescence detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-06-24)
Joshua Ogony, Suneetha Mare, Wei Wu, Nuran Ercal
ABSTRACT
2-Mercaptoethylamine (cysteamine) is an aminothiol compound used as a drug for the treatment of cystinosis, an autosomal recessive lysosomal storage disorder. Because of cysteamine's important role in clinical settings, its analysis by sensitive techniques has become pivotal. Unfortunately, the available methods are either complex or labor intensive. Therefore, we have developed a new rapid, sensitive, and simple method for determining cysteamine in biological samples (brain, kidney, liver, and plasma), using N-(1-pyrenyl) maleimide (NPM) as the derivatizing agent and reversed-phase high performance liquid chromatography (HPLC) with a fluorescence detection method (lambda(ex)=330 nm, lambda(em)=376 nm). The mobile phase was acetonitrile and water (70:30) with acetic acid and o-phosphoric acid (1 mL/L). The calibration curve for cysteamine in serine borate buffer (SBB) was found to be linear over a range of 0-1200 nM (r(2)=0.9993), and in plasma and liver matrix, the r(2) values were 0.9968 and 0.9965, respectively. The coefficients of the variation for the within-run and between-run precisions ranged from 0.68 to 9.90% and 0.63 to 4.17%, respectively. The percentage of relative recovery ranged from 94.1 to 98.6%.
Sigma-Aldrich
N-(1-Pyrenyl)maleimide