MilliporeSigma
  • Palladium-catalyzed DYKAT of butadiene monoepoxide: enantioselective total synthesis of (+)-DMDP, (-)-bulgecinine, and (+)-broussonetine G.
Palladium-catalyzed DYKAT of butadiene monoepoxide: enantioselective total synthesis of (+)-DMDP, (-)-bulgecinine, and (+)-broussonetine G.
Chemistry (Weinheim an der Bergstrasse, Germany) (2006-06-30)
Barry M Trost, Daniel B Horne, Michael J Woltering
ABSTRACT
Palladium catalyzed asymmetric allylic alkylation reaction of an amine with two equivalents of butadiene monoxide allows for the expedient synthesis of trans- and cis-2,5-dihydropyrroles. The versatility of these chiral synthons towards the synthesis of a wide variety of iminosugar natural products was demonstrated with the short and high yielding asymmetric syntheses of (+)-DMDP, and (-)-bulgecinine. In addition, the first total synthesis of (+)-broussonetine G, a potent glycosidase inhibitor, is described along with the assignment of its relative and absolute stereochemical configuration.
Sigma-Aldrich
3,4-Epoxy-1-butene, 98%