MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Spectral and thermal studies of divalent transition metal with indole-2-carboxylic acid and 4-substituted hydrazinethiocarbamide.
Spectral and thermal studies of divalent transition metal with indole-2-carboxylic acid and 4-substituted hydrazinethiocarbamide.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-08-01)
Iman T Ahmed
ABSTRACT
Ternary complexes of Co(II), Ni(II) and Cu(II) with indole-2-carboxylic acid (A) and 4-substituted hydrazinethiocarbamide (L) [4-phenylhydrazinethiocarbamide (L(1)), 4-benzylhydrazinethiocarbamide (L(2)) and 4-(2-propenyl)hydrazinethiocarb-amide (L(3)) were prepared. The structure of the complexes was characterized by microchemical analysis, molar conductance, electronic, IR, (1)H-NMR, mass spectra as well as thermogravimetric studies. An octahedral structure is suggested for Co(II), Ni(II) and Cu(II) ternary complexes.
Sigma-Aldrich
Indole-2-carboxylic acid, 98%