MilliporeSigma
Allergic contact dermatitis from mercapto compounds.
Dermatitis : contact, atopic, occupational, drug (2006-09-08)
Alison K Adams, Erin M Warshaw
ABSTRACT
Despite being well-known allergens, mercaptobenzothiazole and its derivatives continue to be widely used in natural and synthetic rubber, in the mining industry, and in a variety of nonrubber products. The purpose of this article is to review the epidemiology and the clinical characteristics of allergic contact dermatitis from mercapto compounds.
Sigma-Aldrich
2,2′-Dithiobis(benzothiazole), 99%