MilliporeSigma
  • Involvement of Raf-1/MEK/ERK1/2 signaling pathway in zinc-induced injury in rat renal cortical slices.
Involvement of Raf-1/MEK/ERK1/2 signaling pathway in zinc-induced injury in rat renal cortical slices.
The Journal of toxicological sciences (2006-09-09)
Yuka Kohda, Yoshiko Matsunaga, Ryugo Shiota, Tomohiko Satoh, Yuko Kishi, Yoshiko Kawai, Munekazu Gemba
ABSTRACT
Zinc is an essential nutrient that can also be toxic. We have previously reported that zinc-related renal toxicity is due, in part, to free radical generation in the renal epithelial cell line, LLC-PK(1) cells. We have also shown that an MEK1/2 inhibitor, U0126, markedly inhibits zinc-induced renal cell injury. In this study, we investigated the role of an upstream MEK/ERK pathway, Raf-1 kinase pathway, and the transcription factor and ERK substrate Elk-1, in rat renal cortical slices exposed to zinc. Immediately after preparing slices from rat renal cortex, the slices were incubated in medium containing Raf-1 and MEK inhibitors. ERK1/2 and Elk-1 activation were determined by Western blot analysis for phosphorylated ERK (pERK) 1/2 and phosphorylated Elk-1 (pElk-1) in nuclear fractions prepared from slices exposed to zinc. Zinc caused not only increases in 4-hydroxynonenal (4-HNE) modified protein and lipid peroxidation, as an index of oxidant stress, and decreases in PAH accumulation, as that of renal cell injury in the slices. Zinc also induced a rapid increase in ERK/Elk-1 activity accompanied by increased expressions of pERK and pElk-1 in the nuclear fraction. A Raf-1 kinase inhibitor and an MEK1/2 inhibitor U0126 significantly attenuated zinc-induced decreases PAH accumulation in the slices. The Raf-1 kinase inhibitor and U0126 also suppressed ERK1/2 activation in nuclear fractions prepared from slices treated with zinc. The present results suggest that a Raf-1/MEK/ERK1/2 pathway and the ERK substrate Elk-1 are involved in free radical-induced injury in rat renal cortical slices exposed to zinc.
Sigma-Aldrich
N,N′-Diphenyl-p-phenylenediamine, 98%