MilliporeSigma
  • The role of altered acetyl-CoA carboxylase in conferring resistance to fenoxaprop-P-ethyl in Chinese sprangletop (Leptochloa chinensis (L.) Nees).
The role of altered acetyl-CoA carboxylase in conferring resistance to fenoxaprop-P-ethyl in Chinese sprangletop (Leptochloa chinensis (L.) Nees).
Pest management science (2006-09-12)
Tosapon Pornprom, Parnuwat Mahatamnuchoke, Kenji Usui
ABSTRACT
From paddy field observations in 2002 and 2004, fenoxaprop-P-ethyl resistance in Chinese sprangletop (Leptochloa chinensis (L.) Nees) has been studied using information collected from 11 sites in the Saphan-Sung district of Bangkok, Thailand. The resistant Chinese sprangletop was found in nine rice fields, whereas the susceptible Chinese sprangletop was found in only two rice fields. In greenhouse experiments, both fenoxaprop-P-ethyl-resistant and susceptible Chinese sprangletop from the same location were investigated for 50% growth reduction based on phytotoxicity, plant height and fresh and dry weight. The resistant Chinese sprangletop showed apparent resistance at 14-21 days after herbicide application at a rate of 21.1-337.6 g AI ha(-1). The resistance index of resistant Chinese sprangletop was 10-25 times higher than that of the susceptible Chinese sprangletop. In addition, Chinese sprangletop did not exhibit multiple resistance to oxadiazon, propanil and quinclorac. According to acetyl-CoA carboxylase (ACCase) assays, the level of ACCase specific activity in the resistant Chinese sprangletop was significantly higher than that in the susceptible Chinese sprangletop. Similarly, the ACCase activity of the resistant Chinese sprangletop was 10 times less sensitive to fenoxaprop-P-ethyl than that of the susceptible Chinese sprangletop, based on the I50 values. The present study of the mechanism responsible for resistance in the biotypes investigated indicated that there was a close association between the concentration-response at the whole-plant level and ACCase sensitivity to fenoxaprop-P-ethyl, and resistance to fenoxaprop-P-ethyl was conferred by a modified ACCase at the target site, as suggested by higher specific activity and less sensitivity to the herbicide.
Product Description
Supelco
Fenoxaprop-ethyl, PESTANAL®, analytical standard