MilliporeSigma
  • The antibiotic kasugamycin mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
The antibiotic kasugamycin mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.
Nature structural & molecular biology (2006-09-26)
Frank Schluenzen, Chie Takemoto, Daniel N Wilson, Tatsuya Kaminishi, Joerg M Harms, Kyoko Hanawa-Suetsugu, Witold Szaflarski, Masahito Kawazoe, Mikako Shirouzu, Mikako Shirouzo, Knud H Nierhaus, Shigeyuki Yokoyama, Paola Fucini
ABSTRACT
Kasugamycin (Ksg) specifically inhibits translation initiation of canonical but not of leaderless messenger RNAs. Ksg inhibition is thought to occur by direct competition with initiator transfer RNA. The 3.35-A structure of Ksg bound to the 30S ribosomal subunit presented here provides a structural description of two Ksg-binding sites as well as a basis for understanding Ksg resistance. Notably, neither binding position overlaps with P-site tRNA; instead, Ksg mimics codon nucleotides at the P and E sites by binding within the path of the mRNA. Coupled with biochemical experiments, our results suggest that Ksg indirectly inhibits P-site tRNA binding through perturbation of the mRNA-tRNA codon-anticodon interaction during 30S canonical initiation. In contrast, for 70S-type initiation on leaderless mRNA, the overlap between mRNA and Ksg is reduced and the binding of tRNA is further stabilized by the presence of the 50S subunit, minimizing Ksg efficacy.
Sigma-Aldrich
Kasugamycin hydrochloride from Streptomyces kasugaensis