MilliporeSigma
  • Isolation and characterization of Streptomyces sp. NL15-2K capable of degrading lignin-related aromatic compounds.
Isolation and characterization of Streptomyces sp. NL15-2K capable of degrading lignin-related aromatic compounds.
Journal of bioscience and bioengineering (2006-10-10)
Motohiro Nishimura, Osamu Ooi, Julian Davies
ABSTRACT
Strain NL15-2K was isolated from soil by screening for bacteria capable of catabolizing lignin-related aromatic acids. This isolate was identified as a Streptomyces sp. on the basis of morphology and an analysis of its 16S rRNA gene sequence. NL15-2K utilized caffeic acid, coniferyl alcohol, ferulic acid, isovanillic acid, protocatechuic acid, vanillic acid, vanillin, and veratric acid as sole carbon sources.
Sigma-Aldrich
3,4-Dimethoxybenzoic acid, ≥99%