MilliporeSigma
  • Influence of pH and soil copper on adsorption of metalaxyl and penconazole by the surface layer of vineyard soils.
Influence of pH and soil copper on adsorption of metalaxyl and penconazole by the surface layer of vineyard soils.
Journal of agricultural and food chemistry (2006-10-13)
M Arias, M Paradelo, E López, J Simal-Gándara
ABSTRACT
The upper horizons of old vineyard soils have substantial copper contents due to the traditional use of copper-based fungicides. Total copper levels in eight vineyard soils in the Rías Baixas area of Galicia (northwestern Spain) ranged from 60 to 560 mg kg(-1) (mean +/- SD = 206 +/- 170 mg kg(-1)). The adsorption of the fungicides metalaxyl (pK(a) = 1.41) and penconazole (pK(a) = 2.83) by these soils was determined using fungicide solutions of pH 2.5 and 5.5, and desorption of fungicide adsorbed at pH 5.5 was also determined. In all cases, Freundlich equations were fitted to the data with R (2) > 0.96. Penconazole was adsorbed and retained more strongly than metalaxyl, with K(F) values more than an order of magnitude greater. In the desorption experiments, both fungicides exhibited hysteresis. Soil copper content hardly affected the adsorption of metalaxyl, but K(F) values for adsorption of penconazole increased at a rate of about 0.1 mL(n) (microg of penconazole)(1-n) (microg of Cu)(-1), which is attributed to the formation of Cu(2+)-penconazole complexes with greater affinity for soil colloids than penconazole itself. Because the dependence of K(F) for penconazole adsorption on copper content was the same at both pH values, complex formation appears not to have been affected by the solubilization of 6-17% of soil copper at pH 2.5. A similar copper dependence, or lack of dependence, was observed when 100-1000 mg kg(-1) of copper was added as Cu(NO(3))(2).2H(2)O to the solutions from which the fungicides were adsorbed.
Product Description
Supelco
Penconazol, PESTANAL®, analytical standard