MilliporeSigma
  • Relating olfactory neurotoxicity to altered olfactory-mediated behaviors in rainbow trout exposed to three currently-used pesticides.
Relating olfactory neurotoxicity to altered olfactory-mediated behaviors in rainbow trout exposed to three currently-used pesticides.
Aquatic toxicology (Amsterdam, Netherlands) (2006-12-06)
Keith B Tierney, Christopher R Singh, Peter S Ross, Christopher J Kennedy
ABSTRACT
Odor-evoked neurophysiological responses can form the basis for behavioral responses. Here we first characterized olfactory-mediated behavioral and neurophysiological responses of juvenile rainbow trout to the amino acid l-histidine, then looked at whether there were similar responses to the carbamate antisapstain IPBC and the herbicides atrazine and Roundup, and lastly explored how exposures to these pesticides modified the l-histidine responses. Trout were behaviorally attracted to 10(-7)M l-histidine (as assayed in a counter-current olfactometer), but this preference behavior switched to indifference with higher histidine concentrations. Neurophysiologically, the summed electrical responses of peripheral olfactory neurons, as measured using electro-olfactogram (EOG), was 0.843+/-0.252 mV to 10(-7)M l-histidine. Of the pesticides, only Roundup evoked EOGs, indicating the amino acid-based pesticide may have acted as an odorant, and generated a behavioral response: it was avoided at active ingredient [AI; glyphosate isopropyl amine] concentrations > or =10 mg/l. With 30 min pesticide exposures, 10(-7)M l-histidine preference behavior was eliminated following exposure to 1 microg/l IPBC and atrazine, and 100 microg/l AI Roundup. Similarly, 10(-7)M l-histidine-evoked EOGs were significantly reduced by exposure to 1 microg/l IPBC, 10 microg/l atrazine, and 100 microg/l AI Roundup. When combined together, the results demonstrate that typical preference behavior can be abolished when neurophysiological responses are reduced by >60% of control. This asymmetry in response thresholds suggests that behavioral responses may be more sensitive toxicological endpoints than neurophysiological responses.
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%