MilliporeSigma
  • Facile construction of lanthanide metallomacrocycles with the bridging imidazolate and triazolate ligands and their ring expansions.
Facile construction of lanthanide metallomacrocycles with the bridging imidazolate and triazolate ligands and their ring expansions.
Inorganic chemistry (2007-01-03)
Jie Zhang, Ruifang Cai, Zhenxia Chen, Xigeng Zhou
ABSTRACT
Four novel tri- or tetranuclear organolanthanide metallomacrocycles [Cp2Ln(mu-Im)(THF)3 (Cp = C5H5, Ln = Yb (1), Er (2)], [Cp2Dy(mu-Im)]4(THF)]3 x 2THF (3), and [Cp'2Yb(mu-eta1:eta2-Tz)]4 x 2THF (Cp' = CH3C5H4) (4) have been synthesized through protolysis of Cp3Ln or Cp'3Yb with imidazole or triazole, indicating that both the bridge-ligand size and the lanthanide-ion radii can be applied in the modulation of the metallomacrocycles. Further investigations on the reactivity of complexes 1, 3, and 4 toward phenyl isocyanate reveal that PhNCO inserts readily into the simple bridge Ln-N bonds of 1 and 3 to yield the corresponding insertion products [Cp2Ln(mu-eta1:eta2-OC(Im)NPh)]3 (Ln = Yb (5), Dy (6)) but cannot insert into the Ln-N bond with a mu-eta1:eta2-bonding mode in 4. The novel bridge ligand [OC(Im)NPh] can expand the numbers of the ring members from 12 to 18 in 5 or 16 to 18 in 6. The number of metal atoms in the metallacycles with the ligand [OC(Im)NPh] is independent of the lanthanide-ion size; both trinuclear lanthanide macrocycles are observed in 5 and 6. All of these new complexes have been characterized by elemental analysis and spectroscopic properties, and their structures have also been determined through X-ray single-crystal diffraction analysis.
Sigma-Aldrich
Phenyl isocyanate, ≥98%
Supelco
Phenyl isocyanate, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)