MilliporeSigma
  • Functional characterization of Musca glutamate- and GABA-gated chloride channels expressed independently and coexpressed in Xenopus oocytes.
Functional characterization of Musca glutamate- and GABA-gated chloride channels expressed independently and coexpressed in Xenopus oocytes.
Insect molecular biology (2007-01-05)
Y Eguchi, M Ihara, E Ochi, Y Shibata, K Matsuda, S Fushiki, H Sugama, Y Hamasaki, H Niwa, M Wada, F Ozoe, Y Ozoe
ABSTRACT
Ligand-gated chloride channels (LGICs) are important targets for insecticides and parasiticides. Genes encoding subunits of two LGICs, a glutamate-gated chloride channel (MdGluCl-alpha) and a gamma-aminobutyric acid (GABA)-gated chloride channel (MdRdl), were cloned from house-flies (Musca domestica L.). These genes were first expressed independently in Xenopus laevis oocytes by cRNA injection in order to investigate the pharmacology of these ligand-gated channels using two-electrode voltage-clamp electrophysiology. It was found that L-glutamate and GABA activated the MdGluCl-alpha homo-oligomers with an EC(50) value of 30 microM and the MdRdl homo-oligomers with an EC(50) value of 101 microM, respectively. Both channels were chloride ion-permeable, and the MdRdl channel was more sensitive to chloride channel blockers, such as gamma-hexachlorocyclohexane (gamma-HCH), fipronil and picrotoxinin, than the MdGluCl-alpha channel. MdGluCl-alpha required only 1-2 days of incubation after cRNA injection to be expressed in oocytes, whereas 4-7 days of incubation was necessary to achieve MdRdl expression. However, when the cRNA of MdGluCl-alpha was injected at a dose of 1% (w/w) 1 day after the injection of the cRNA of MdRdl, a significant increase in the current amplitude of responses to GABA was observed, and the incubation period necessary for MdRdl expression became shorter. These results suggest that MdGluCl-alpha assists in the expression of MdRdl when the two are coexpressed.
Sigma-Aldrich
Picrotoxinin